Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMC2 SIRNA Assays ›
Gene Symbol:
SMC2
Gene Name:
structural maintenance of chromosomes 2
Gene Aliases:
CAP-E, CAPE, SMC-2, SMC2L1
Chromosome Location:
Chr.9: 104093808 - 104141419 on Build GRCh38
UniGene ID:
Hs.119023
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
18122
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042550.1
NP_001036015.1
23
3475
NM_001042551.1
NP_001036016.1
23
3413
NM_001265602.1
NP_001252531.1
23
3429
NM_006444.2
NP_006435.2
23
3404
XM_006716933.3
XP_006716996.1
23
3326
XM_011518148.2
XP_011516450.1
23
3289
XM_011518149.2
XP_011516451.1
23
3331
XM_011518150.2
XP_011516452.1
22
3193
XM_011518151.2
XP_011516453.1
22
3171
XM_011518152.2
XP_011516454.1
23
3355
XM_011518153.1
XP_011516455.1
23
3355
XM_017014206.1
XP_016869695.1
23
3475
XM_017014207.1
XP_016869696.1
23
3377
XM_017014208.1
XP_016869697.1
23
3417
XM_017014209.1
XP_016869698.1
22
3304
XM_017014210.1
XP_016869699.1
22
3194
XM_017014211.1
XP_016869700.1
22
3283
XM_017014212.1
XP_016869701.1
22
3150
GenBank mRNA
AF092563.1
-
23
3339
AF113673.1
-
4
409
AJ420415.1
-
6
431
AK000955.1
-
12
1646
AK001485.1
-
23
3461
AK292599.1
-
23
3352
AL833191.1
-
23
3414
BC130385.1
-
23
3259
40146416
BC144163.1
-
23
3275
9052683
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitotic chromosome condensation
meiotic chromosome condensation
meiotic chromosome segregation
cell division
kinetochore organization
protein binding
ATP binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-s7sbm:80/100.66.79.121:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline