Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144522.5 | NP_653105.3 | 8 | 1814 | ||
GenBank mRNA | AF285112.1 | - | 8 | 1172 | ||
AK047320.1 | - | 8 | 1815 | |||
AK047327.1 | - | 8 | 1815 | |||
BC025889.1 | - | 2 | 207 | |||
BC072576.1 | - | 8 | 1357 | 6401332 |