Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144522.5 | NP_653105.3 | 9 | 3554 | ||
GenBank mRNA | AK047320.1 | - | 9 | 3555 | ||
AK047327.1 | - | 9 | 3555 |