Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144524.2 | NP_653107.2 | 10 | 3284 | ||
GenBank mRNA | AK003979.1 | - | 1 | 172 | ||
AK143549.1 | - | 10 | 3289 | |||
BC019148.1 | - | 10 | 3298 | 5052974 |