Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144829.1 | NP_659078.1 | 7 | 764 | ||
GenBank mRNA | AK136332.1 | - | 7 | 731 | ||
AK168281.1 | - | 7 | 731 | |||
BC005711.1 | - | 7 | 764 | 3497962 |