Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164566.1 | NP_001158038.1 | 6 | 753 | ||
NM_144882.4 | NP_659131.2 | 8 | 939 | |||
GenBank mRNA | AK031266.1 | - | 8 | 894 | ||
AK032169.1 | - | 8 | 874 | |||
BC014764.1 | - | 6 | 753 | 4217648 | ||
BC021879.1 | - | 8 | 875 | 4216816 | ||
CK624383.1 | - | 1 | 46 | |||
CN698528.1 | - | 2 | 290 |