Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145131.1 | NP_660113.1 | 5 | 447 | ||
GenBank mRNA | AF513714.1 | - | 5 | 447 | ||
AK129292.1 | - | 5 | 421 | |||
AK141277.1 | - | 5 | 442 | |||
AK147837.1 | - | 5 | 438 | |||
AK164235.1 | - | 5 | 409 | |||
AK167426.1 | - | 5 | 438 | |||
AK168335.1 | - | 5 | 438 | |||
AY779273.1 | - | 5 | 447 | |||
AY779274.1 | - | 5 | 447 |