Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114131.1 | NP_001107603.1 | 20 | 3566 | ||
NM_146081.2 | NP_666193.2 | 20 | 3617 | |||
GenBank mRNA | AK013951.2 | - | 7 | 1619 | ||
AK133969.1 | - | 20 | 3490 | |||
AK167713.1 | - | 19 | 3028 | |||
BC020371.1 | - | 8 | 1701 | |||
BC026489.1 | - | 12 | 2590 | 4504305 | ||
BC029761.1 | - | 20 | 3552 | 3989055 |