Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146171.1 | NP_666283.1 | 5 | 523 | ||
GenBank mRNA | AK129068.1 | - | 5 | 488 | ||
AK156143.1 | - | 5 | 553 | |||
BC025460.1 | - | 5 | 543 | |||
BC026982.1 | - | 5 | 523 | 5364933 | ||
BC029133.1 | - | 5 | 480 | 4951596 |