Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf113a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153503.1 | NP_705723.1 | 1 | 756 | ||
GenBank mRNA | AK013178.1 | - | 1 | 817 | ||
AK146283.1 | - | 1 | 855 | |||
BC024906.1 | - | 1 | 756 | 3495576 |