Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172397.3 | NP_765985.1 | 5 | 2500 | ||
GenBank mRNA | AK032430.1 | - | 5 | 2501 | ||
AK040486.1 | - | 5 | 2502 | |||
AK049809.1 | - | 5 | 2470 | |||
AK167349.1 | - | 5 | 2470 | |||
BC068130.1 | - | 5 | 2444 | 6413206 |