Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172400.3 | NP_765988.2 | 5 | 351 | ||
GenBank mRNA | AK003892.2 | - | 5 | 340 | ||
AK049526.1 | - | 4 | 878 | |||
AK088325.1 | - | 5 | 355 | |||
AK150933.1 | - | 5 | 346 | |||
AK168645.1 | - | 5 | 335 | |||
BC035539.1 | - | 5 | 310 | |||
BC057191.1 | - | 5 | 322 | |||
BC064732.1 | - | 5 | 330 | |||
BC066222.1 | - | 5 | 330 | 30459247 | ||
BC080760.1 | - | 4 | 864 | |||
BC137735.1 | - | 5 | 347 | 8860754 | ||
BC145655.1 | - | 5 | 347 | |||
BY754536.1 | - | 5 | 355 |