Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167767.1 | NP_001161239.1 | 4 | 516 | ||
NM_001167768.1 | NP_001161240.1 | 4 | 505 | |||
NM_172401.4 | NP_765989.3 | 4 | 524 | |||
GenBank mRNA | AI746795.1 | - | 4 | 516 | ||
AK079099.1 | - | 4 | 508 | |||
AK162914.1 | - | 4 | 486 | |||
BC046986.1 | - | 3 | 443 | 6476504 | ||
BC057311.1 | - | 4 | 496 | 6811681 |