Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175118.3 | NP_780327.1 | 2 | 548 | ||
GenBank mRNA | AK002934.1 | - | 2 | 547 | ||
AK088940.1 | - | 2 | 495 | |||
AK163560.1 | - | 2 | 557 | |||
BC051598.1 | - | 2 | 525 | |||
BC119127.1 | - | 2 | 537 | 8733876 |