Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175118.3 | NP_780327.1 | 2 | 1208 | ||
GenBank mRNA | AK002934.1 | - | 2 | 1207 | ||
AK088940.1 | - | 2 | 1152 | |||
AK163560.1 | - | 2 | 1217 | |||
AW493803.1 | - | 1 | 216 |