Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175121.3 | NP_780330.2 | 4 | 591 | ||
GenBank mRNA | AK018767.1 | - | 2 | 430 | ||
AK030917.1 | - | 3 | 745 | |||
AK031615.1 | - | 4 | 622 | |||
AK047403.1 | - | 3 | 769 | |||
AK129342.1 | - | 4 | 552 | |||
AK164568.1 | - | 2 | 318 | |||
BC027259.1 | - | 4 | 495 | |||
BC041108.1 | - | 4 | 461 | 4038546 | ||
BC048178.1 | - | 4 | 591 | 6825801 |