Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100626.1 | NP_001094096.1 | 6 | 523 | ||
NM_006459.3 | NP_006450.2 | 5 | 767 | |||
XM_005269442.3 | XP_005269499.1 | 6 | 553 | |||
XM_017015541.1 | XP_016871030.1 | 6 | 537 | |||
GenBank mRNA | AF064093.1 | - | 6 | 499 | ||
AK056574.1 | - | 5 | 3124 | |||
AK222480.1 | - | 6 | 495 | |||
AK298427.1 | - | 5 | 413 | |||
AK315580.1 | - | 6 | 495 | |||
BC031791.1 | - | 5 | 436 | 4214940 |