Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175124.5 | NP_780333.3 | 10 | 2494 | ||
GenBank mRNA | AK077270.1 | - | 10 | 2490 | ||
AK159417.1 | - | 10 | 2464 | |||
BC027058.1 | - | 7 | 2052 | |||
BC032971.1 | - | 1 | 253 | |||
BC086694.1 | - | 10 | 2385 | 30697866 |