Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175126.4 | NP_780335.1 | 1 | 1629 | ||
GenBank mRNA | AK053859.1 | - | 1 | 1366 | ||
BC055755.1 | - | 1 | 1611 | 6412435 |