Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175127.2 | NP_780336.1 | 5 | 1085 | ||
GenBank mRNA | AK076211.1 | - | 5 | 1091 | ||
AK122295.1 | - | 5 | 1070 | |||
AK162091.1 | - | 5 | 1055 | |||
AK162188.1 | - | 5 | 966 | |||
BC055458.1 | - | 4 | 787 | |||
BC078447.1 | - | 2 | 933 | |||
BC094542.1 | - | 5 | 1085 | 6408744 |