Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930430F08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175128.2 | NP_780337.2 | 5 | 736 | ||
GenBank mRNA | AK015251.1 | - | 5 | 685 | ||
AK016270.1 | - | 568 | ||||
AK032226.1 | - | 5 | 692 | |||
AK036647.1 | - | 5 | 719 | |||
AK077761.1 | - | 5 | 731 | |||
AK078231.1 | - | 5 | 737 | |||
AK082894.1 | - | 5 | 689 | |||
AK160693.1 | - | 5 | 719 | |||
BC082587.1 | - | 5 | 686 | 30630155 |