Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175140.4 | NP_780349.3 | 4 | 1642 | ||
GenBank mRNA | AB106878.1 | - | 3 | 1240 | ||
AF308145.1 | - | 4 | 1460 | |||
AK005217.1 | - | 4 | 1376 | |||
AK038952.1 | - | 3 | 1615 | |||
AK051290.1 | - | 4 | 1224 | |||
BC051124.1 | - | 4 | 1607 | 6528978 |