Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175151.4 | NP_780360.1 | 12 | 991 | ||
GenBank mRNA | AK042248.1 | - | 12 | 992 |