Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53inp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178111.3 | NP_835212.1 | 3 | 535 | ||
GenBank mRNA | AJ297793.1 | - | 3 | 467 | ||
AK146589.1 | - | 3 | 540 | |||
AK159265.1 | - | 1 | 111 | |||
AK170132.1 | - | 3 | 573 | |||
BC036958.1 | - | 2 | 342 | 4504446 | ||
BC043086.1 | - | 3 | 535 | 6417598 |