Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 3, 4 | 373 | ||
GenBank mRNA | AK003990.1 | - | 3, 4 | 329 | ||
AK039949.1 | - | 3, 4 | 373 | |||
AK041783.1 | - | 367 | ||||
AK084395.1 | - | 3, 4 | 367 | |||
AK086633.1 | - | 365 | ||||
AK144975.1 | - | 3, 4 | 374 | |||
AK159740.1 | - | 3, 4 | 385 | |||
AK161378.1 | - | 3, 4 | 363 | |||
AK167806.1 | - | 3, 4 | 367 | |||
AK168510.1 | - | 3, 4 | 329 | |||
AK169066.1 | - | 3, 4 | 363 | |||
AK169351.1 | - | 3, 4 | 330 | |||
BC119178.1 | - | 3, 4 | 367 | 8733927 | ||
BC119180.1 | - | 3, 4 | 367 | 8733929 |