Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135934.1 | NP_001129406.1 | 21 | 2935 | ||
NM_001135935.1 | NP_001129407.1 | 21 | 2941 | |||
NM_001135936.1 | NP_001129408.1 | 20 | 2851 | |||
NM_001286665.1 | NP_001273594.1 | 22 | 3025 | |||
NM_001286666.1 | NP_001273595.1 | 20 | 2845 | |||
NM_001286667.1 | NP_001273596.1 | 19 | 2761 | |||
NM_006475.2 | NP_006466.2 | 23 | 3106 | |||
XM_005266231.2 | XP_005266288.1 | 22 | 3017 | |||
XM_005266232.2 | XP_005266289.1 | 22 | 3011 | |||
GenBank mRNA | AY918092.1 | - | 20 | 2744 | ||
BG938993.1 | - | 2 | 548 | |||
D13665.1 | - | 21 | 2860 | |||
D13666.1 | - | 23 | 2999 |