Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178616.3 | NP_848731.2 | 12 | 1138 | ||
GenBank mRNA | AK007547.1 | - | 12 | 1107 | ||
AK012951.1 | - | 11 | 1602 | |||
AK029365.1 | - | 8 | 709 | |||
AK051738.1 | - | 12 | 1128 | |||
AK076002.1 | - | 12 | 1126 | |||
AK089942.1 | - | 12 | 1103 | |||
AK136049.1 | - | 12 | 1101 | |||
AK140773.1 | - | 1123 | ||||
AK167559.1 | - | 12 | 1109 | |||
AK168289.1 | - | 12 | 1130 | |||
BC030432.1 | - | 9 | 792 | |||
BC037590.1 | - | 12 | 1087 | |||
BC055457.1 | - | 12 | 1070 | |||
BC090664.1 | - | 12 | 1090 | 30677672 | ||
BC090980.1 | - | 12 | 1105 | 30283128 | ||
BC100595.1 | - | 12 | 1100 | 6767671 | ||
BC119136.1 | - | 12 | 1109 | 8733885 | ||
BC119138.1 | - | 12 | 1109 | 8733887 |