Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183271.2 | NP_899094.1 | 2 | 255 | ||
GenBank mRNA | AK006775.1 | - | 2 | 249 | ||
AK018854.1 | - | 2 | 249 | |||
AK018888.1 | - | 2 | 256 | |||
AK018909.1 | - | 2 | 227 | |||
BC100423.1 | - | 2 | 234 | 30919644 |