Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1500015L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005254.1 | - | 3 | 345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_183277.1 | - | 345 | Chr. - - [-] on Build |