Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197982.3 | NP_932099.2 | 2 | 335 | ||
GenBank mRNA | AK011285.1 | - | 1 | 175 | ||
AK012002.1 | - | 2 | 245 | |||
AK016960.1 | - | 2 | 245 | |||
AK137114.1 | - | 241 | ||||
AK145927.1 | - | 2 | 278 | |||
AK146294.1 | - | 2 | 280 | |||
AK149475.1 | - | 2 | 246 | |||
AK150997.1 | - | 2 | 264 | |||
AK151395.1 | - | 2 | 245 | |||
AK152394.1 | - | 2 | 245 | |||
AK162752.1 | - | 803 | ||||
AK164289.1 | - | 2 | 205 | |||
AK167890.1 | - | 2 | 205 | |||
AK168079.1 | - | 2 | 204 | |||
AK172448.1 | - | 2 | 264 | |||
BC020134.1 | - | 2 | 223 | 3969048 | ||
BY065389.1 | - | 2 | 314 | |||
CF172497.1 | - | 2 | 232 |