Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam58b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197989.1 | NP_932106.1 | 1 | 239 | ||
GenBank mRNA | AK007413.1 | - | 1 | 187 | ||
AK032408.1 | - | 193 | ||||
BC027022.1 | - | 1 | 239 | 5367363 |