Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197993.2 | NP_932110.1 | 2 | 215 | ||
GenBank mRNA | AK044659.1 | - | 1 | 1597 | ||
AK138418.1 | - | 2 | 135 | |||
BC023900.1 | - | 2 | 192 | 5321265 | ||
BC025499.1 | - | 2 | 239 | 5320398 |