Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019507.2 | NP_062380.2 | 2, 3 | 845 | ||
GenBank mRNA | AF093099.1 | - | 2, 3 | 803 | ||
AF241242.1 | - | 2, 3 | 835 | |||
AK054495.1 | - | 2, 3 | 844 | |||
AK156271.1 | - | 2, 3 | 846 | |||
AY836551.1 | - | 1, 2 | 158 | |||
BC137986.1 | - | 2, 3 | 763 | 8861005 | ||
BC137988.1 | - | 2, 3 | 763 | 8861007 |