Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019507.2 | NP_062380.2 | 3 | 850 | ||
GenBank mRNA | AF093099.1 | - | 3 | 808 | ||
AF241242.1 | - | 3 | 840 | |||
AK054495.1 | - | 3 | 849 | |||
AK156271.1 | - | 3 | 851 | |||
AY836551.1 | - | 2 | 163 | |||
BC137986.1 | - | 3 | 768 | 8861005 | ||
BC137988.1 | - | 3 | 768 | 8861007 |