Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123041.2 | NP_001116513.2 | 2 | 1082 | ||
NM_001123396.1 | NP_001116868.1 | 2 | 1082 | |||
XM_011534069.1 | XP_011532371.1 | 3 | 795 | |||
GenBank mRNA | AK292685.1 | - | 2 | 1092 | ||
AK292920.1 | - | 2 | 700 | |||
BC074751.2 | - | 1 | 636 | 30915210 | ||
BC095540.1 | - | 1 | 601 | 7001971 | ||
BC126452.1 | - | 1 | 631 | 8992168 | ||
BC136396.1 | - | 1 | 631 | 9020383 | ||
D29984.1 | - | 1 | 597 | |||
U03882.1 | - | 1 | 636 | |||
U03905.1 | - | 2 | 677 |