Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289514.1 | NP_001276443.1 | 17 | 2735 | ||
NM_019636.3 | NP_062610.2 | 18 | 3316 | |||
GenBank mRNA | AK122445.1 | - | 20 | 4404 | ||
AK148356.1 | - | 17 | 2692 | |||
AK154591.1 | - | 19 | 3352 | |||
AK164923.1 | - | 14 | 2020 | |||
AK168763.1 | - | 7 | 1021 | |||
BC004675.1 | - | 7 | 974 | |||
BC059824.1 | - | 18 | 3490 | |||
BC117931.1 | - | 17 | 2984 | 40097712 | ||
U33005.1 | - | 18 | 3272 |