Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019800.4 | NP_062774.2 | 4, 5 | 934 | ||
GenBank mRNA | AB030038.1 | - | 4, 5 | 927 | ||
AF216223.1 | - | 4, 5 | 411 | |||
AK002783.1 | - | 4, 5 | 848 | |||
AK077729.1 | - | 4, 5 | 922 | |||
AK160271.1 | - | 4, 5 | 853 | |||
AK165356.1 | - | 4, 5 | 935 | |||
AK171757.1 | - | 4, 5 | 849 | |||
BC047276.1 | - | 4, 5 | 674 | 6487551 |