Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019811.3 | NP_062785.2 | 3 | 518 | ||
GenBank mRNA | AF216873.1 | - | 3 | 468 | ||
AK028586.1 | - | 3 | 496 | |||
AK029014.1 | - | 3 | 486 | |||
AK029262.1 | - | 504 | ||||
AK053877.1 | - | 3 | 492 | |||
AK086921.1 | - | 3 | 491 | |||
AK093831.1 | - | 3 | 495 | |||
BC040790.1 | - | 3 | 468 | |||
BC051432.1 | - | 3 | 518 | 6515568 |