Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167604.1 | NP_001161076.1 | 13 | 1320 | ||
NM_001324128.1 | NP_001311057.1 | 11 | 1167 | |||
NM_001324131.1 | NP_001311060.1 | 13 | 1364 | |||
NM_001324132.1 | NP_001311061.1 | 12 | 1231 | |||
NM_001324133.1 | NP_001311062.1 | 13 | 1320 | |||
NM_001324134.1 | NP_001311063.1 | 11 | 1151 | |||
NM_001324135.1 | NP_001311064.1 | 12 | 1269 | |||
NM_001324136.1 | NP_001311065.1 | 13 | 1535 | |||
NM_020383.3 | NP_065116.3 | 13 | 1320 | |||
XM_011540134.1 | XP_011538436.1 | 13 | 1314 | |||
XM_017016613.1 | XP_016872102.1 | 12 | 1170 | |||
XM_017016614.1 | XP_016872103.1 | 14 | 1303 | |||
XM_017016615.1 | XP_016872104.1 | 12 | 1187 | |||
GenBank mRNA | AF195530.1 | - | 12 | 1147 | ||
AF272981.1 | - | 11 | 1071 | |||
AK093693.1 | - | 11 | 1125 | |||
AK223513.1 | - | 12 | 1131 | |||
AK289436.1 | - | 13 | 1223 | |||
AK295756.1 | - | 5 | 576 | |||
AK304367.1 | - | 13 | 1213 | |||
AL833411.1 | - | 13 | 1226 | |||
BC005126.2 | - | 12 | 1109 | 3836436 | ||
BC007579.1 | - | 13 | 1211 | 3139868 | ||
BC013417.2 | - | 9 | 821 | |||
BX537682.1 | - | 10 | 976 | |||
CR456922.1 | - | 11 | 1071 | |||
X95762.1 | - | 11 | 1071 |