Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 6 | 825 | ||
NM_001252490.1 | NP_001239419.1 | 5 | 699 | |||
NM_001252491.1 | NP_001239420.1 | 4 | 905 | |||
NM_001252492.1 | NP_001239421.1 | 5 | 1068 | |||
NM_001252493.1 | NP_001239422.1 | 5 | 1068 | |||
NM_207530.3 | NP_997413.2 | 19 | 1954 | |||
GenBank mRNA | AB017026.1 | - | 3 | 949 | ||
AF394063.2 | - | 3 | 260 | |||
AK050641.1 | - | 5 | 623 | |||
AK132676.1 | - | 6 | 824 | |||
AK133806.1 | - | 3 | 331 | |||
AK161374.1 | - | 7 | 696 | |||
AK168366.1 | - | 5 | 1029 | |||
AY536214.1 | - | 19 | 1833 | |||
BB667982.1 | - | 1 | 27 | |||
BC015075.1 | - | 5 | 789 | |||
BC046466.1 | - | 5 | 831 | |||
BC057194.1 | - | 4 | 332 | |||
BC076637.1 | - | 5 | 699 | |||
BC092247.1 | - | 4 | 905 | |||
DV068702.1 | - | 3 | 295 |