Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 9 | 1272 | ||
NM_001252490.1 | NP_001239419.1 | 8 | 1146 | |||
NM_001252491.1 | NP_001239420.1 | 7 | 1352 | |||
NM_001252492.1 | NP_001239421.1 | 8 | 1515 | |||
NM_001252493.1 | NP_001239422.1 | 8 | 1515 | |||
NM_207530.3 | NP_997413.2 | 22 | 2401 | |||
GenBank mRNA | AB017026.1 | - | 6 | 1396 | ||
AK050641.1 | - | 8 | 1070 | |||
AK132676.1 | - | 9 | 1271 | |||
AK133806.1 | - | 6 | 778 | |||
AK168366.1 | - | 8 | 1476 | |||
AY536214.1 | - | 22 | 2280 | |||
BB667982.1 | - | 4 | 474 | |||
BC076637.1 | - | 8 | 1146 | |||
BC092247.1 | - | 7 | 1352 | |||
CO426253.1 | - | 2 | 268 | |||
DV068702.1 | - | 6 | 742 |