Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020575.2 | NP_065600.1 | 3 | 421 | ||
GenBank mRNA | AF155739.1 | - | 3 | 421 | ||
AK034109.1 | - | 446 | ||||
AK143177.1 | - | 3 | 441 | |||
AK166666.1 | - | 3 | 428 | |||
AK167246.1 | - | 3 | 403 | |||
AK168555.1 | - | 3 | 424 | |||
BC025029.1 | - | 3 | 385 | 4935124 |