Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020578.3 | NP_065603.2 | 1 | 475 | ||
GenBank mRNA | AF155883.1 | - | 1 | 446 | ||
AF506002.1 | - | 1 | 270 | |||
AK051717.1 | - | 1 | 366 | |||
AK128964.1 | - | 1 | 454 | |||
AK141265.1 | - | 1 | 477 | |||
BC046596.1 | - | 1 | 219 | 6312263 |