Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271003.1 | NP_001257932.1 | 2, 3 | 551 | ||
NM_001271004.1 | NP_001257933.1 | 2, 3 | 584 | |||
NM_006528.3 | NP_006519.1 | 2, 3 | 584 | |||
GenBank mRNA | AK024817.1 | - | 2, 3 | 346 | ||
AK075427.1 | - | 2, 3 | 345 | |||
AK092499.1 | - | 2, 3 | 313 | |||
AK129833.1 | - | 2, 3 | 346 | |||
AK313260.1 | - | 2, 3 | 346 | |||
AY691946.1 | - | 2, 3 | 322 | |||
BC005330.1 | - | 2, 3 | 346 | 3929933 | ||
BC034155.1 | - | 1, 2 | 69 | |||
D29992.1 | - | 2, 3 | 327 | |||
DA831865.1 | - | 2, 3 | 584 | |||
DA833200.1 | - | 2, 3 | 345 | |||
EF488667.1 | - | 185 | ||||
L27624.1 | - | 2, 3 | 309 |