Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113527.1 | NP_001106999.1 | 3 | 724 | ||
NM_001291220.1 | NP_001278149.1 | 4 | 1042 | |||
NM_001291221.1 | NP_001278150.1 | 4 | 708 | |||
NM_020583.5 | NP_065608.2 | 4 | 915 | |||
GenBank mRNA | AF217484.1 | - | 4 | 1008 | ||
AK005528.1 | - | 4 | 709 | |||
AK008139.1 | - | 3 | 725 | |||
AK008171.1 | - | 3 | 725 | |||
AK008353.1 | - | 3 | 727 | |||
AK136756.1 | - | 4 | 841 | |||
BC022751.1 | - | 4 | 862 | 4912320 |