Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020595.2 | NP_065620.1 | 3, 4 | 364 | ||
GenBank mRNA | AF243504.1 | - | 3, 4 | 364 | ||
AJ243939.1 | - | 3, 4 | 379 | |||
BC139152.1 | - | 3, 4 | 463 | 8862174 | ||
BC139153.1 | - | 3, 4 | 463 | 8862175 |