Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020619.2 | NP_065644.2 | 3 | 747 | ||
GenBank mRNA | AK144946.1 | - | 3 | 748 | ||
AK157286.1 | - | 3 | 744 | |||
BC051949.1 | - | 3 | 747 | 5716189 |