Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021311.3 | NP_067286.1 | 9 | 1107 | ||
GenBank mRNA | AB032604.1 | - | 9 | 1072 | ||
AF438405.1 | - | 9 | 1107 | |||
BC066846.1 | - | 1158 | ||||
BC129857.1 | - | 8 | 957 | 40109791 | ||
BC129858.1 | - | 8 | 957 | 40109793 | ||
EF196092.1 | - | 7 | 917 |