Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF171060.1 | - | 10 | 946 | ||
AK002399.1 | - | 10 | 967 | |||
AK171752.1 | - | 10 | 944 | |||
BC022715.1 | - | 10 | 932 | |||
BC037100.1 | - | 10 | 954 | |||
BC048821.1 | - | 10 | 954 | 3966642 | ||
BE853767.1 | - | 4 | 439 |