Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021315.2 | NP_067290.2 | 17, 18 | 2090 | ||
GenBank mRNA | AB077991.1 | - | 17, 18 | 2053 | ||
AF233884.1 | - | 6, 7 | 553 | |||
AK134051.1 | - | 17, 18 | 2049 | |||
AK168944.1 | - | 17, 18 | 2093 | |||
BC031132.1 | - | 17, 18 | 2053 | 4954951 |